Gfp (from spinal cord)
: Biorbyt
- Source:Spinal cord
- Storage conditions:Lyophilized GFP although stable at room temperature for weeks, should be stored desiccated below −18 °C. Upon reconstitution GFP should be stored at 4 °C between 2-7 days and for future use below −18 °C. For long term storage it is recommended to add carrier protein (0. 1% HSA or BSA). Please prevent freeze/thaw cycles.
- Protein synonyms:Glial Filament
- Protein/peptide name:GFP
- Purity:Greater than 98.0% as determined by RP-HPLC and analysis by SDS-PAGE
- Solubility:It is recommended to reconstitute the lyophilized GFP in sterile H2O not less than 100ug/ml, which can then be further diluted to other aqueous solutions.
- Formulation:The protein was lyophilized from 1mg/ml solution containing 10mM sodium phosphate buffer pH 7.5, 6M urea, 2mM DTT, 1mM EDTA and 10mM methylammonium chloride.
- Tested applications:SDS-PAGE
Frequently Bought Together