Human FABP4 (from E. coli)
Supplier: Biorbyt
Certificates
About this item
Specifications
- Source:E. coli
- Species:Human
- Storage conditions:Store lyophilized protein at −20 °C. Aliquot the product after reconstitution to avoid repeated freezing/thawing cycles. Reconstituted protein can be stored at 4 °C for limited period of time; it does not show any change after two weeks at 4 °C. The lyophilized protein remains stable until the expiry date when stored at −20 °C.
- Protein synonyms:Fatty acid-binding
- Protein/peptide name:FABP4
- Purity:Greater than 90% as determined by SDS-PAGE. Two-step procedure using size exclusion chromatography before and after refolding.
- Solubility:0.1M Acetate buffer pH4 and let the lyophilized pellet dissolve completely. For conversion into higher pH value, we recommend intensive dilution by relevant buffer to concentration of 10ug/ml. In higher concentrations the solubility of this antigen is lim
- Formulation:Sterile filtered and lyophilized from 0.5 mg/ml in 0.05M Acetate buffer pH4.
- Tested applications:SDS-PAGE
Specifications
Frequently Bought Together