Order Entry
Hungary
ContactUsLinkComponent
Human recombinant ADFP (from E. coli)
Human recombinant ADFP (from E. coli)
Supplier:  Biorbyt
Certificates
Human recombinant ADFP (from E. coli)
Supplier:  Biorbyt

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    Store lyophilized protein at −20 °C. Aliquot the product after reconstitution to avoid repeated freezing/thawing cycles. Reconstituted protein can be stored at 4 °C for limited period of time; it does not show any change after two weeks at 4 °C. The lyophilized protein remains stable until the expiry date when stored at −20 °C.
  • Protein synonyms:
    Adipophilin
  • Protein/peptide name:
    ADFP
  • Purity:
    Greater than 95% as determined by SDS PAGE.
  • Solubility:
    Add deionized water to prepare working stock solution of approximately 0.5mg/mL and let the lyophilized pellet dissolve completely. Product is not sterile! Please filter the product by an appropriate sterile filter before using it in the cell culture.
  • Sequence:
    MKHHHHHHAS VAVDPQPSVV TRVVNLPLVS STYDLMSSAY LSTKDQYPYL KSVCEMAENG VKTITSVAMT SALPIIQKLE PQIAVANTYA CKGLDRIEER LPILNQPSTQ IVANAKGAVT GAKDAVTTTV TGAKDSVAST ITGVMDKTKG AVTGSVEKTK SVVSGSINTV LGSRMMQLVS SGVENALTKS ELLVEQYLPL TEEELEKEAK KVEGFDLVQK PSYYVRLGSL STKLHSRAYQ QALSRVKEAK QKSQQTISQL HSTVHLIEFA RKNVYSANQK IQDAQDKLYL SWVEWKRSIG YDDTDESHCA EHIESRTLAI ARNLTQQLQT TCHTLLSNIQ GVPQNIQDQA KHMGVMAGDI YSVFRNAASF KEVSDSLLTS SKGQLQKMKE SLDDVMDYLV NNTPLNWLVG PFYPQLTESQ NAQDQGAEMD KSSQETQRSEHKTH.
  • Formulation:
    Human ADFP was lyophilized from 0.5mg/ml solution containing 20 mM Tris pH-7.5, and 20 mM NaCl.
  • Tested applications:
    SDS-PAGE

Specifications

Related Information

Frequently Bought Together