Order Entry
Hungary
ContactUsLinkComponent
Human recombinant REG1B (from E. coli)
Human recombinant REG1B (from E. coli)
Supplier:  Biorbyt
Certificates
Human recombinant REG1B (from E. coli)
Supplier:  Biorbyt

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    Store lyophilized protein at −20 °C. Aliquot the product after reconstitution to avoid repeated freezing/thawing cycles. Reconstituted protein can be stored at 4 °C for limited period of time; it does not show any change after two weeks at 4 °C.The lyophilized protein remains stable until the expiry date when stored at −20 °C.
  • Protein synonyms:
    Lithostathine-1-beta
  • Protein/peptide name:
    REG1B
  • Purity:
    Greater than 95% as determined by SDS-PAGE.
  • Solubility:
    It is recommended to add deionized water to working concentration approximately 0.5 mg/ml and let the lyophilized pellet dissolve completely. Product is not sterile! Please filter the product by appropriate sterile filter before using it in the cell cultu
  • Formulation:
    Filtered (0.4 um) and lyophilized from 0.5 mg/ml in 20 mM Tris, pH 8.0.
  • Tested applications:
    SDS-PAGE

Specifications

Related Information

Frequently Bought Together