Human recombinant PTGDS (from E. coli)
: Biorbyt
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:Lyophilized PTGDS although stable at room temperature for weeks, should be stored desiccated below −18 °C. Upon reconstitution PTGDS should be stored at 4 °C between 2-7 days and for future use below −18 °C. For long term storage it is recommended to add carrier protein (0. 1% HSA or BSA). Please prevent freeze/thaw cycles.
- Protein synonyms:Prostaglandin-H2 D-isomerase
- Protein/peptide name:PTGDS
- Purity:Greater than 95.0% as determined by RP-HPLC and analysis by SDS-PAGE
- Solubility:It is recommended to reconstitute the lyophilized PTGDS in sterile H2O not less than 100ug/ml, which can then be further diluted to other aqueous solutions.
- Sequence:MRGSHHHHHH GMASAPEAQV SVQPNFQQDK FLGRWFSAGL ASNSSWLREK KAALSMCKSV VAPATDGGLN LTSTFLRKNQ CETRTMLLQP AGSLGSYSYR SPHWGSTYSV SVVETDYDQY ALLYSQGSKG PGEDFRMATL YSRTQTPRAE LKEKFTAFCK AQGFTEDTIV FLPQTDKCMT EQ.
- Formulation:Filtered (0.4 umicron) and lyophilized from 0.5 mg/ml in 20 mM Tris buffer, 20 mM NaCl, pH 7.5.
- Tested applications:SDS-PAGE
Frequently Bought Together