Human Jun kinase 2, alpha 2 (from E. coli)
Supplier: Biorbyt
Certificates
About this item
Specifications
- Source:E. coli
- Species:Human
- Storage conditions:Aliquot and store at −20 °C or −80 °C. Avoid repeated freeze/thaw cycles.
- Protein synonyms:Mitogen-activated
- Protein/peptide name:Jun kinase 2, alpha 2
- Purity:Greater than 95% as determined by SDS-PAGE.
- Formulation:The protein concentration is 0.32mg/ml in 50mM Tris, pH 7.5, 0.15M NaCl, 0.2 um sucrose, 10mM beta mercaptoethanol, 1mM EDTA and 0.1% Triton X-100.
- Tested applications:SDS-PAGE
Specifications
Frequently Bought Together