Order Entry
Hungary
ContactUsLinkComponent
 
Human CK2A1 (from E. coli)
 :  Biorbyt
 
Human CK2A1 (from E. coli)
 :  Biorbyt

 

  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    Aliquot and store at −20 °C or −80 °C. Avoid repeated freeze/thaw cycles.
  • Protein synonyms:
    EC 27111
  • Protein/peptide name:
    CK2A1
  • Purity:
    Greater than 90% as determined by SDS-PAGE.
  • Sequence:
    MGSSHHHHHH SSGLVPRGSH MSGPVPSRAR VYTDVNTHRP REYWDYESHV VEWGNQDDYQLVRKLGRGKY SEVFEAINIT NNEKVVVKIL KPVKKKKIKR EIKILENLRG GPNIITLADI VKDPVSRTPA LVFEHVNNTD FKQLYQTLTD YDIRFYMYEI LKALDYCHSM GIMHRDVKPH NVMIDHEHRK LRLIDWGLAE FYHPGQEYNV RVASRYFKGP ELLVDYQMYD YSLDMWSLGC MLASMIFRKE PFFHGHDNYD QLVRIAKVLG TEDLYDYIDK YNIELDPRFN DILGRHSRKRWERFVHSENQ HLVSPEALDF LDKLLRYDHQ SRLTAREAME HPYFYTVVKD QARMGSSSMP GGSTPVSSAN MMSGISSVPT PSPLGPLAGS PVIAAANPLG MPVPAAAGAQ Q.
  • Formulation:
    CSNK2A1 is supplied in 20 mM Tris pH-8.0, 500mM NaCl, 1mM DTT and 50% glycerol.
  • Tested applications:
    SDS-PAGE

 

 

Frequently Bought Together