Order Entry
Hungary
ContactUsLinkComponent
Human CK beta (from E. coli)
Human CK beta (from E. coli)
Supplier:  Biorbyt
Certificates
Human CK beta (from E. coli)
Supplier:  Biorbyt

Specifications

  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    Store at 4 °C if entire vial will be used within 2-4 weeks. Store frozen at −20 °C for longer periods of time. For long term storage it is recommended to add carrier protein (0. 1% HSA or BSA). Please avoid freeze/thaw cycles.
  • Protein synonyms:
    CK II beta
  • Protein/peptide name:
    CK beta
  • Purity:
    Greater than 95.0% as determinedAnalysis by SDS-PAGE.
  • Sequence:
    MSSSEEVSWI SWFCGLRGNE FFCEVDEDYI QDKFNLTGLN EQVPHYRQAL DMILDLEPDE ELEDNPNQSD LIEQAAEMLY GLIHARYILT NRGIAQMLEK YQQGDFGYCP RVYCENQPML PIGLSDIPGE AMVKLYCPKC MDVYTPKSSR HHHTDGAYFG TGFPHMLFMV HPEYRPKRPA NQFVPRLYGF KIHPMAYQLQ LQAASNFKSP VKTIR.
  • Formulation:
    20 mM Tris pH 8.0, 200mM NaCl, 1mM DTT, 1mM EDTA, 1uM leupeptin, 40% glycerol.
  • Tested applications:
    SDS-PAGE , HPLC , FuncS

Specifications

Related Information

Frequently Bought Together