Mouse recombinant mIL 22 PEG (from E. coli)
Supplier: Biorbyt
Certificates
About this item
Specifications
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Mouse
- Storage conditions:Lyophilized pegylated murine IL22 although stable at room temperature for several days, should be stored desiccated below −20 °C. Upon reconstitution at 0. 1mg/ml pegylated mouse IL22 and up to 2mg/ml, filter and sterilized, the protein can be stored at °C Celsius for several weeks making it suitable for long term infusion studies using osmotic pumps. At lower concentration addition of carrier protein (0. 1% HSA or BSA) is suggested. Please prevent freeze/thaw cycles.
- Biological activity:The ED50 as determined by STAT3 phosphorylation assay in HepG cells. The activity in vitro was found to be 10% compared to the non-pegylated mouse IL22.
- Protein synonyms:IL-TIF
- Protein/peptide name:mIL 22 PEG
- Purity:Greater than 98.0% as determined by: (a) Analysis by Gel-Filtration. (b) Analysis by SDS-PAGE.
- Solubility:It is recommended to reconstitute the lyophilized pegylated mouse Interleukin -22 in sterile 0.4% NaHCO3 adjusted to pH 8-9 not less than 100ug/ml, which can then be further diluted to other aqueous solutions.
- Formulation:Lyophilized from concentrated solution at 0.65mg/ml containing 0.003mM NaHCO3.
- Tested applications:SDS-PAGE
Specifications
Frequently Bought Together