Mouse VEGF (from E. coli)
Supplier: Biorbyt
Certificates
About this item
Specifications
- Source:E. coli
- Species:Mouse
- Storage conditions:Mouse VEGF although stable 4 °C for weeks, should be stored desiccated below −18 °C. For long term storage it is recommended to add carrier protein (0. 1% HSA or BSA). Please prevent freeze/thaw cycles.
- Protein synonyms:Vascular endothelial growth factor A
- Protein/peptide name:VEGF
- Purity:Greater than 90.0% as determined by SDS-PAGE.
- Sequence:MGSSHHHHHH SSGLVPRGSH MAPTTEGEQK SHEVIKFMDV YQRSYCRPIE TLVDIFQEYP DEIEYIFKPS CVPLMRCAGC CNDEALECVP TSESNITMQI MRIKPHQSQH IGEMSFLQHS RCECRPKKDR TKPEKCDKPR R.
- Formulation:The Mouse VEGF contains 20 mM Tris-HCl buffer (pH8.0) and 10% glycerol.
- Tested applications:SDS-PAGE
Specifications
Frequently Bought Together