Dog recombinant apoliprotein HEK (from HEK293 cells)
Supplier: Biorbyt
Certificates
About this item
Specifications
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Dog
- Storage conditions:Store lyophilized protein at −20 °C. Aliquot the product after reconstitution to avoid repeated freezing/thawing cycles. Reconstituted protein can be stored at 4 °C for limited period of time; it does not show any change after two weeks at 4 °C. The lyophilized protein remains stable until the expiry date when stored at −20 °C.
- Protein synonyms:AAG4
- Protein/peptide name:Apoliprotein HEK
- Purity:Greater than 95% as determined by SDS PAGE.
- Solubility:Add deionized H2O to working volume of 0.5mg/ml and let the lyophilized pellet dissolve completely.
- Sequence:PGDYKDDDDK PAGDQAVSDT ELQEMSTEGS KYINKEIKNA LKGVKQIKTL IEQTNEERKS LLSNLEEAKK KKEDALNDTK DSETKLKASQ GVCNDTMMAL WEECKPCLKQ TCMKFYARVC RSGSGLVGHQ LEEFLNQSSP FYFWMNGDRI DSLLENDRQQ THALDVMQDS FNRASSIMDE LFQDRFFTRE PQDTYHYSPF SLFQRRPFFN PKFRIARNII PFPRFQPLNF HDMFQPFFDM IHQAQQAMDV NLHRIPYHFP IEFPEEDNRT VCKEIRHNST GCLKMKDQCE KCQEILSVDC SSNNPAQVQL RQELSNSLQI AEKFTKLYDE LLQSYQEKMF NTSSLLKQLN EQFSWVSQLA NLTQSEDPFY LQVTTVGSQT SDSNVPVGFT KVVVKLFDSD PITVMIPEAV SRNNPKFMET VAEKALQEYRQKHREE.
- Formulation:Canine Clusterin was lyophilized from 0.5mg/ml solution containing 20 mM Tris buffer and 20 mM NaCl, pH7.5.
- Tested applications:SDS-PAGE
Specifications
Frequently Bought Together