Recombinant RANTES (from E. coli)
Supplier: Biorbyt
Certificates
About this item
Specifications
- Protein/peptide type:Recombinant
- Source:E. coli
- Storage conditions:Store at 4 °C if entire vial will be used within 2-4 weeks. Store frozen at −20 °C for longer periods of time. Please avoid freeze thaw cycles.
- Protein synonyms:Beta chemokine RANTES
- Protein/peptide name:RANTES
- Purity:Greater than 95.0% as determined by SDS-PAGE.
- Formulation:Rantes His-Tag protein is supplied in 1x PBS and 50% glycerol.
- Tested applications:SDS-PAGE
Specifications
Frequently Bought Together