Order Entry
Hungary
ContactUsLinkComponent
Treponema p41, mosaic
Treponema p41, mosaic
Supplier:  Biorbyt
Certificates
Treponema p41, mosaic
Supplier:  Biorbyt

Specifications

  • Storage conditions:
    Five years frozen. One month in solution at room temperature.
  • Protein/peptide name:
    Treponema p41 Mosaic
  • Purity:
    Treponema Pallidum protein is >90% pure as determined by 10% PAGE (coomassie staining). Treponema Pallidum protein was purified by proprietary chromatographic technique.
  • Formulation:
    25mM Tris-HCl pH-8, 60mM NaCl 50% glycerol.
  • Tested applications:
    SDS-PAGE

Specifications

Related Information

Frequently Bought Together