HIV-1 gp41 L-Biotin (from E. coli)
Supplier: Biorbyt
Certificates
About this item
Specifications
- Source:E. coli
- Storage conditions:HIV-1 gp41 Long is shipped at ambient temperature. Upon arrival, Store at −20 °C Stable for Five years frozen. One month in solution at room temperature.
- Protein/peptide name:HIV-1 gp41 L-Biotin
- Purity:Greater than 95.0% as determined by HPLC analysis SDS-PAGE.
- Formulation:0.01M Na2CO3, 0.01M Na3EDTA, 0.014 beta mercaptoethanol and 0.05% Tween 20.
- Tested applications:SDS-PAGE
Specifications
Frequently Bought Together