HCV NS5 Genotype-1a
Supplier: Biorbyt
Certificates
About this item
Specifications
- Storage conditions:Five years frozen. One month in solution at room temperature.
- Protein/peptide name:HCV NS5 Genotype-1a
- Purity:HCV NS5 Genotype-1a protein is >95% pure as determined by 10% PAGE (coomassie staining). HCV NS5 Genotype-1a protein was purified by proprietary chromatographic technique.
- Formulation:50mM Tris. pH & 5mM EDTA.
- Tested applications:SDS-PAGE
Specifications
Frequently Bought Together