Recombinant Der-F1
Supplier: Biorbyt
Certificates
About this item
Specifications
- Protein/peptide type:Recombinant
- Storage conditions:Five years frozen. One month in solution at room temperature.
- Protein synonyms:Peptidase 1
- Protein/peptide name:Der-F1
- Purity:Protein is >95% pure as determined by 10% PAGE (coomassie staining) and RP-HPLC.
- Formulation:10mM Tris-HCl, pH 6.5, 100mM Sodium Phosphate and 8M urea.
- Tested applications:SDS-PAGE
Specifications
Frequently Bought Together