Order Entry
Hungary
ContactUsLinkComponent
Recombinant PA2-BVP
Recombinant PA2-BVP
Supplier:  Biorbyt
Certificates
Recombinant PA2-BVP
Supplier:  Biorbyt

Specifications

  • Protein/peptide type:
    Recombinant
  • Storage conditions:
    Five years frozen. One month in solution at room temperature.
  • Protein synonyms:
    Phospholipase A2
  • Protein/peptide name:
    PA2-BVP
  • Purity:
    Protein is >95% pure as determined by 10% PAGE (coomassie staining) and RP-HPLC.
  • Formulation:
    10mM Tris-HCl, pH 6.5, 0.1M Sodium Phosphate and 8M urea.
  • Tested applications:
    SDS-PAGE

Specifications

Related Information

Frequently Bought Together