Recombinant p53
Catalog # BIRBORB98853-100
Supplier: Biorbyt
CAS Number:
Specifications
- Pk:100 µG
- Protein/peptide type:Recombinant
- Storage conditions:Store at −20 °C. Avoid multiple freeze/thaw cycles.
- Protein synonyms:BCC7
- Protein/peptide name:p53
- Purity:Greater than 95% pure SDS PAGE non-reducing, Electrophoresis
- Molecular weight:53 kDa
- Formulation:Liquid, 20mM Tris-HCL,150Mm NaCl, pH 8.0
- Tested applications:SDS-PAGE