HTLV-1 p24 Core
Catalog # BIRBORB85275-100
Supplier: Biorbyt
CAS Number:
Specifications
- Pk:100 µG
- Conjugation:Unconjugated
- Storage conditions:Store at−20 °C.
- Protein/peptide name:HTLV-1 p24 Core
- Purity:Greater than 95% by SDS-PAGE and RP-HPLC
- Formulation:Liquid. Supplied in 50 mM NaPO4 pH 6.0, 1 mM DTTand 1 mM EDTA.
- Tested applications:WB , SDS-PAGE