Order Entry
Hungary
ContactUsLinkComponent
HCV-NS3-S2b
HCV-NS3-S2b
  BIRBORB97915-100
 :  Biorbyt
 :  
HCV-NS3-S2b
  BIRBORB97915-100
 :  Biorbyt
 :  

 

  • Pk:
    100 µG
  • Storage conditions:
    Store at−20 °C.
  • Protein/peptide name:
    HCV-NS3-S2b
  • Purity:
    Greater than 95% by SDS-PAGE and RP-HPLC
  • Formulation:
    Liquid. Supplied in 25 mM Tris-HCl pH 7.5, 1 mM EDTA,1.5 M urea and 50% glycerol.
  • Tested applications:
    ELISA , WB