HCV-NS3-S2c
Catalog # BIRBORB97913-100
Supplier: Biorbyt
CAS Number:
Specifications
- Pk:100 µG
- Storage conditions:Store at−20 °C.
- Protein/peptide name:HCV-NS3-S2c
- Purity:Greater than 95% by SDS-PAGE
- Formulation:Liquid. Supplied in 1.5M urea, 25mM Tris-HCl pH 8,0.2% Triton-X and 50% glycerol.
- Tested applications:ELISA , WB