Rat recombinant rTpc1808 (from E. coli)
Supplier: Biorbyt
Certificates
About this item
Specifications
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Rat
- Storage conditions:Lyophilized Tpc1808 although stable 10 °C for week, should be stored desiccated below −18 °C. Please prevent freeze/thaw cycles.
- Protein synonyms:Tropic 1808
- Protein/peptide name:rTpc1808
- Purity:Greater than 95.0% as determined by RP-HPLC and analysis by SDS-PAGE
- Sequence:MSYYHHHHHHMNLAQIAALNQISNLNAIRVGQVLKVSNAAGSNNTQNTTQPSAGVPTNTASSTTGYTVKSGDTLSAIAAANGVSLANLLSWNNLSLQAIIYPGQKLTIQNANNATVTTPNAPTSTPTVMPSTNGSYTVKSGDTLYGIAAKLGTNVQTLLSLNGLQLSSTIYVGQVLKTTGAVAGAGTATSTPTPVTPTVSKPAAANGVSTAGLSAAQAAWLRTAVVDAQAATAGTGVLASVTVAQAILESGWGQSALASAPYHNFNLYLIKVKNTWKLMTLLLS.
- Formulation:The Tropic-1808 was lyophilized from 1X PBS, pH 7.4.
- Tested applications:SDS-PAGE
Specifications
Frequently Bought Together