Human Recombinant CD14-CHO (from CHO cells)
Gyártó: Biorbyt
Tanúsítványok
Termék információk
Specifikációk
- Protein/peptide type:Recombinant
- Source:CHO cells
- Species:Human
- Storage conditions:Lyophilized CD14 although stable at room temperature for weeks, should be stored desiccated below −18 °C. Upon reconstitution CD14 should be stored at 4 °C between 2-7 days and for future use below −18 °C. For long term storage it is recommended to add carrier protein (0. 1% HSA or BSA). Please prevent freeze/thaw cycles.
- Biological activity:Up to 20 ug/ml CD14 inhibit binding of FITC-LPS (0.5ug/ml) to x 105 CD14C HO transfectants (FACS).
- Protein synonyms:Monocyte differentiation antigen CD14
- Protein/peptide name:CD14-CHO
- Purity:Greater than 95.0% as determined by SDS-PAGE.
- Solubility:It is recommended to reconstitute the lyophilized CD14 in sterile H2O not less than 100ug/ml. Further dilutions should be made with phosphate buffered saline (PBS).
- Formulation:CD14 was lyophilized from concentrated protein solution (1.0 mg/ml) containing phosphate-buffered saline, pH 7.2.
- Tested applications:SDS-PAGE
Specifikációk
Frequently Bought Together