Order Entry
Hungary
ContactUsLinkComponent
Human SUMO1 (from E. coli)
Human SUMO1 (from E. coli)
Supplier:  Biorbyt
Certificates
Human SUMO1 (from E. coli)
Supplier:  Biorbyt

Specifications

  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    SUMO1 although stable at 4 °C for week, should be stored desiccated below −18 °C. For long term storage it is recommended to add carrier protein (0.1% HSA or BSA).Please prevent freeze-thaw cycles.
  • Protein synonyms:
    Small ubiquitin-related modifier 1
  • Protein/peptide name:
    SUMO1
  • Purity:
    Greater than 98.0% as determined by RP-HPLC and analysis by SDS-PAGE
  • Formulation:
    10mM sodium chloride, 100mM imidazole, 0.5mM PMSF, 1mM DTT and 10% glycerol.
  • Tested applications:
    SDS-PAGE

Specifications

Related Information

Frequently Bought Together