Human IL6 (from E. coli)
Supplier: Biorbyt
Certificates
About this item
Specifications
- Source:E. coli
- Species:Human
- Storage conditions:Store at 4 °C if entire vial will be used within 2-4 weeks. Store frozen at −20 °C for longer periods of time. Please avoid freeze/thaw cycles.
- Protein synonyms:Interleukin BSF 2
- Protein/peptide name:IL6
- Purity:Greater than 95.0% as determined by RP-HPLC and analysis by SDS-PAGE
- Formulation:Interleukin-6 His-Tag protein is supplied in 10mM Tris HCl pH 8.0 and 50% glycerol.
- Tested applications:SDS-PAGE
Specifications
Frequently Bought Together