Mouse adiponectin (Globular) (from E. coli)
Supplier: Biorbyt
Certificates
About this item
Specifications
- Source:E. coli
- Species:Mouse
- Storage conditions:Liquid Acrp30 Mouse although stable at 4 °C for week, should be stored below −18 °C. Please prevent freeze/thaw cycles.
- Gene ID:9370
- Protein synonyms:ACDC
- Protein/peptide name:Adiponectin (globular)
- Purity:Greater than 95.0% as determined by SDS-PAGE.
- Sequence:MAYMYRSAFS VGLETRVTVP NVPIRFTKIF YNQQNHYDGS TGKFYCNIPG LYYFSYHITVYMKDVKVSLF KKDKAVLFTY DQYQEKNVDQ ASGSVLLHLE VGDQVWLQVY GDGDHNGLYADNVNDSTFTG FLLYHDTN.
- Formulation:Acrp30 Mouse (1mg/ml) solution containing 20 mM Tris-HCl pH7.5, 50mM NaCl, 5mM DTT and 10% Glycerol.
- Tested applications:SDS-PAGE
Specifications
Frequently Bought Together