Recombinant SPA 41kDa (from E. coli)
: Biorbyt
- Protein/peptide type:Recombinant
- Source:E. coli
- Storage conditions:Protein-A should be stored at all times at 4 °C.
- Biological activity:7mg human IgG/mg protein-A.
- Protein synonyms:Immunoglobulin G-binding
- Protein/peptide name:SPA 41kDa
- Purity:Greater than 98.0% as determined by SDS-PAGE RP-HPLC.
- Formulation:The protein was lyophilized from concentrated (1mg/ml) solution containing no additives.
- Tested applications:SDS-PAGE
Frequently Bought Together