Human recombinant JNK2 (from E. coli)
: Biorbyt
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:Aliquot and store at −20 °C or −80 °C. Avoid repeated freeze/thaw cycles.
- Protein synonyms:Mitogen-activated kinase 9
- Protein/peptide name:JNK2
- Purity:Greater than 95% as determined by SDS-PAGE.
- Formulation:JNK2/SAPK1 (1.3 mg/ml) is supplied in 50mM Tris-HCl, 100mM NaCl, 1mM DTT, 50% glycerol, pH 8.5.5
- Tested applications:SDS-PAGE
Frequently Bought Together