Order Entry
Hungary
ContactUsLinkComponent
Recombinant PRKACA (from E. coli)
Recombinant PRKACA (from E. coli)
Supplier:  Biorbyt
Certificates
Recombinant PRKACA (from E. coli)
Supplier:  Biorbyt

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Storage conditions:
    Aliquot and store at −20 °C or −80 °C. Avoid repeated freeze/thaw cycles.
  • Protein synonyms:
    cAMP-dependent
  • Protein/peptide name:
    PRKACA
  • Purity:
    Greater than 95% as determined by SDS-PAGE.
  • Formulation:
    PKA catalytic subunit is supplied in buffer containing 25mM potassium phosphate (pH 6.5), 5mM 2-mercaptoethanol, 5mM EDTA, 150mM NaCI and 50% glycerol.
  • Tested applications:
    SDS-PAGE

Specifications

Related Information

Frequently Bought Together