Recombinant PKACa2 (from E. coli)
Supplier: Biorbyt
Certificates
About this item
Specifications
- Protein/peptide type:Recombinant
- Source:E. coli
- Storage conditions:PKA should be stored at 4 °C if entire vial will be used within 2-4 weeks. For long term storage it is recommended to store at −20 °C. Avoid multiple freeze-thaw cycles.
- Biological activity:Holoenzyme can be activated by adding the second messenger cAMP (Activation constant about 100nM) releasing two monomeric catalytic subunits.
- Protein synonyms:Protein Kinase holoenzyme type alpha
- Protein/peptide name:PKACa2
- Purity:Greater than 95.0% as determined by SDS-PAGE.
- Formulation:PKA holoenzyme type-I alpha is supplied in 50% glycerol.
- Tested applications:SDS-PAGE
Specifications
Frequently Bought Together