Order Entry
Hungary
ContactUsLinkComponent
Mouse murine MCP3 (from E. coli)
Mouse murine MCP3 (from E. coli)
Supplier:  Biorbyt
Certificates
Mouse murine MCP3 (from E. coli)
Supplier:  Biorbyt

Specifications

  • Source:
    E. coli
  • Species:
    Mouse
  • Storage conditions:
    Store at 4 °C for up to two weeks. For long term storage, aliquot and store at −20 °C, avoid freeze/thaw cycles.
  • Protein synonyms:
    C-C motif chemokine 7
  • Protein/peptide name:
    Murine MCP3
  • Purity:
    > 95% by SDS-PAGE and HPLC analyses
  • Molecular weight:
    8.5 kDa
  • Sequence:
    QPDGPNASTC CYVKKQKIPK RNLKSYRRIT SSRCPWEAVI FKTKKGMEVCAEAHQKWVEE AIAYLDMKTP TPKP
  • Formulation:
    Lyophilized from a 0.2 um filtered concentrated solution in 2xPBS, pH 7.4.
  • Tested applications:
    SDS-PAGE , HPLC

Specifications

Related Information

Frequently Bought Together