Rhesus macaque S100B (from E. coli)
Supplier: Biorbyt
Certificates
About this item
Specifications
- Source:E. coli
- Species:Rhesus monkey
- Storage conditions:Store at 4 °C for up to two weeks. For long term storage, aliquot and store at −20 °C, avoid freeze/thaw cycles.
- Protein synonyms:Rhesus macaque S100 calcium binding B
- Protein/peptide name:S100B
- Purity:> 97% by SDS-PAGE and HPLC analyses
- Molecular weight:10.7 kDa
- Sequence:SELEKAMVAL IDVFHQYSGR EGDKHKLKKS ELKELINNEL SHFLEEIKEQ EVVDKVMETLDSDGDGECDF QEFMAFVAMV TTACHEFFEH E
- Formulation:Lyophilized from a 0.2 um filtered solution in PBS, pH 7.4
- Tested applications:SDS-PAGE , HPLC
Specifications
Frequently Bought Together