Order Entry
Hungary
ContactUsLinkComponent
 
Mouse PF4 (from E. coli)
 :  Biorbyt
 
Mouse PF4 (from E. coli)
 :  Biorbyt

 

  • Source:
    E. coli
  • Species:
    Mouse
  • Storage conditions:
    Store at 4 °C for up to two weeks. For long term storage, aliquot and store at −20 °C, avoid freeze/thaw cycles.
  • Protein synonyms:
    Murine Platelet Factor-4
  • Protein/peptide name:
    PF4
  • Purity:
    > 97% by SDS-PAGE and HPLC analyses
  • Molecular weight:
    8.2 kDa
  • Sequence:
    VTSAGPEESD GDLSCVCVKT ISSGIHLKHI TSLEVIKAGR HCAVPQLIAT LKNGRKICLDRQAPLYKKVI KKILES
  • Formulation:
    Lyophilized from a 0.2 um filtered concentrated solution in 20mM PB, 1.5M NaCl, pH 7.4.
  • Tested applications:
    SDS-PAGE , HPLC

 

 

Frequently Bought Together