Order Entry
Hungary
ContactUsLinkComponent
Human PDI (from E. coli)
Human PDI (from E. coli)
Supplier:  Biorbyt
Certificates
Human PDI (from E. coli)
Supplier:  Biorbyt

Specifications

  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    Store at 4 °C for up to two weeks. For long term storage, aliquot and store at −20 °C, avoid freeze/thaw cycles.
  • Protein synonyms:
    Protein Disulfide Isomerase
  • Protein/peptide name:
    PDI
  • Purity:
    > 95% by SDS-PAGE and HPLC analyses
  • Molecular weight:
    56.6 kDa
  • Sequence:
    MRGSGSHHHH HHAPEEEDHV LVLRKSNFAE ALAAHKYLLV EFYAPWCGHCKALAPEYAKA AGKLKAEGSE IRLAKVDATE ESDLAQQYGV RGYPTIKFFRNGDTASPKEY TAGREADDIV NWLKKRTGPA ATTLPDGAAA ESLVESSEVAVIGFFKDVES DSAKQFLQAA EAIDDIPFGI TSNSDVFSKY QLDKDGVVLFKKFDEGRNNF EGEVTKENLL DFIKHNQLPL VIEFTEQTAP KIFGGEIKTHILLFLPKSVS DYDGKLSNFK TAAESFKGKI LFIFIDSDHT DNQRILEFFGLKKEECPAVR LITLEEEMTK YKPESEELTA ERITEFCHRF LEGKIKPHLMSQELPEDWDK QPVKVLVGKN FEDVAFDEKK NVFVEFYAPW CGHCKQLAPIWDKLGETYKD HENIVIAKMD STANEVEAVK VHSFPTLKFF PASADRTVIDYNGERTLDGF KKFLESGGQD GAGDDDDLED LEEAEEPDME EDDDQKAVKD EL
  • Formulation:
    Lyophilized from a 0.2 um filtered concentrated solution in PBS, pH 7.0.
  • Tested applications:
    SDS-PAGE , HPLC

Specifications

Related Information

Frequently Bought Together