Human leptin (from E. coli)
Catalog # BIRBORB107774-200
Supplier: Biorbyt
CAS Number:
Specifications
- Pk:200 µG
- Source:E. coli
- Species:Human
- Storage conditions:Store at 4 °C for up to two weeks. For long term storage, aliquot and store at −20 °C, avoid freeze/thaw cycles.
- Protein synonyms:LEP
- Protein/peptide name:Leptin
- Purity:> 97% by SDS-PAGE and HPLC analyses.
- Molecular weight:16.0 kDa
- Sequence:VPIQKVQDDT KTLIKTIVTR INDISHTQSV SSKQKVTGLD FIPGLHPILT LSKMDQTLAVYQQILTSMPS RNVIQISNDL ENLRDLLHVL AFSKSCHLPW ASGLETLDSL GGVLEASGYSTEVVALSRLQ GSLQDMLWQL DLSPGC
- Formulation:Lyophilized from a 0.2 um filtered concentrated solution in PBS, pH 7.4.
- Tested applications:SDS-PAGE , HPLC