Rat GMFB (from E. coli)
: Biorbyt
- Source:E. coli
- Species:Rat
- Storage conditions:Store at 4 °C for up to two weeks. For long term storage, aliquot and store at −20 °C, avoid freeze/thaw cycles.
- Protein synonyms:Glia Maturation Factor beta
- Protein/peptide name:GMFB
- Purity:> 98% by SDS-PAGE and HPLC analyses
- Molecular weight:16.5 KDa
- Sequence:SESLVVCDVA EDLVEKLRKF RFRKETNNAA IIMKIDKDKR LVVLDEELEG ISPDELKDELPERQPRFIVY SYKYQHDDGR VSYPLCFIFS SPVGCKPEQQ MMYAGSKNKL VQTAELTKVFEIRNTEDLTE EWLREKLGFF H
- Formulation:Lyophilized from a 0.2 um filtered concentrated solution in PBS, pH 7.4.
- Tested applications:SDS-PAGE , HPLC
Frequently Bought Together