Order Entry
Hungary
ContactUsLinkComponent
 
Human GDNF (from E. coli)
 :  Biorbyt
 
Human GDNF (from E. coli)
 :  Biorbyt

 

  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    Store at 4 °C for up to two weeks. For long term storage, aliquot and store at −20 °C, avoid freeze/thaw cycles.
  • Protein synonyms:
    Glial Cell-derived Neurotrophic Factor
  • Protein/peptide name:
    GDNF
  • Purity:
    > 97% by SDS-PAGE and HPLC analyses.
  • Molecular weight:
    15.2 kDa
  • Sequence:
    MSPDKQMAVL PRRERNRQAA AANPENSRGK GRRGQRGKNR GCVLTAIHLNVTDLGLGYET KEELIFRYCS GSCDAAETTY DKILKNLSRN RRLVSDKVGQ ACCRPIAFDDDLSFLDDNLV YHILRKHSAK RCGCI
  • Formulation:
    Lyophilized from a 0.2 um filtered concentrated solution in 20mM PB, 500mM NaCl, pH 6.4
  • Tested applications:
    SDS-PAGE , HPLC

 

 

Frequently Bought Together