Order Entry
Hungary
ContactUsLinkComponent
Mouse Cardiotrophin 1 (from E. coli)
Mouse Cardiotrophin 1 (from E. coli)
Supplier:  Biorbyt
Certificates
Mouse Cardiotrophin 1 (from E. coli)
Supplier:  Biorbyt

Specifications

  • Source:
    E. coli
  • Species:
    Mouse
  • Storage conditions:
    Store at 4 °C for up to two weeks. For long term storage, aliquot and store at −20 °C, avoid freeze/thaw cycles.
  • Protein synonyms:
    Murine Cardiotrophin1
  • Protein/peptide name:
    Cardiotrophin 1
  • Purity:
    > 95 % by SDS-PAGE and HPLC analyses
  • Molecular weight:
    18.2 kDa
  • Sequence:
    LRPGDCEVCI SYLGRFYQDL KDRDVTFSPA TIENELIKFC REARGKENRL CYYIGATDDAATKIINEVSK PLAHHIPVEK ICEKLKKKDS QICELKYDKQ IDLSTVDLKK LRVKELKKILDDWGETCKGC AEKSDYIRKI NELMPKYAPK AASARTDL
  • Formulation:
    Lyophilized from a 0.2 um filtered concentrated solution in 20 mM Tris, 300 mM NaCl, 3 mM beta -mercaptoethanol, pH 8.5
  • Tested applications:
    SDS-PAGE , HPLC

Specifications

Related Information

Frequently Bought Together