Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0 Ft
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Hungary
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Proteins and Peptides
HckGST SH3 domain
HckGST SH3 domain
HckGST SH3 domain
Catalog # BIRBORB98112-50
Supplier:
Biorbyt
CAS Number:
Expand image
HckGST SH3 domain
Catalog # BIRBORB98112-50
Supplier:
Biorbyt
CAS Number:
Specifications
Pk:
50 µG
Storage conditions:
−20 °C
UniProtKB:
5HCK
Protein/peptide name:
HckGST SH3 domain
Purity:
90% pure by SDS-PAGE
Formulation:
Lyophilized
Specifications
About this item
Product Details
Documentation
Product Details
Documentation