HSV I Ag (from E. coli)
Catalog # BIRBORB98957-1
Supplier: Biorbyt
CAS Number:
Specifications
- Pk:1 mg
- Source:E. coli
- Storage conditions:Store at −20 °C. Avoid multiple freeze/thaw cycles.
- Protein/peptide name:HSV I Ag
- Purity:Greater than 95% pure SDS PAGE non-reducing, Electrophoresis
- Formulation:PBS, pH 7.4
- Tested applications:ELISA , WB , IHC-P , RIA