Order Entry
Hungary
Orders LinkContactUsLinkComponent
Recombinant FABP Ag

Recombinant FABP Ag

100 µG
Catalog # BIRBORB98925-100
Marketsource
Supplier:   Biorbyt
Recombinant FABP Ag
Catalog # BIRBORB98925-100
Supplier:  Biorbyt
Marketsource
CAS Number:  
Recombinant FABP Ag

100 µG

CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 100 µG
Product Details & Documents

Documents

Specifications

Specifications

  • Pk:
    100 µG
  • Cat. No.:
    BIRBORB98925-100
  • Protein/peptide type:
    Recombinant
  • Protein/peptide name:
    FABP Ag
  • Storage conditions:
    Store at −20 °C. Avoid multiple freeze/thaw cycles.
  • Formulation:
    PBS, pH 7.4
  • Purity:
    Greater than 95% pure SDS PAGE non-reducing, Electrophoresis
  • Tested applications:
    ELISA

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.