Order Entry
Hungary
ContactUsLinkComponent
Recombinant HCV NS4 C100-3
Recombinant HCV NS4 C100-3
Catalog # BIRBORB98870-1
Supplier:  Biorbyt
CAS Number:  
Recombinant HCV NS4 C100-3
Catalog # BIRBORB98870-1
Supplier:  Biorbyt
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Protein/peptide type:
    Recombinant
  • Storage conditions:
    Store at −20 °C. Avoid multiple freeze/thaw cycles.
  • Protein/peptide name:
    HCV NS4 C100-3
  • Purity:
    > 95% Pure (SDS-PAGE)
  • Formulation:
    20 mM Tris-HCl, pH 8.0
  • Tested applications:
    ELISA

Specifications

About this item