Human recombinant Cystatin C
BIRBORB79660-0.1
: Biorbyt
:
- Pk:0,1 mg
- Protein/peptide type:Recombinant
- Species:Human
- Storage conditions:Store at -2 to -8 °C.
- Protein synonyms:AD8
- Protein/peptide name:Cystatin C
- Purity:Greater than 95% pure SDS-PAGE. Protein G Chromatography