Order Entry
Hungary
ContactUsLinkComponent
 
Human recombinant Cystatin C
Catalog # BIRBORB79659-1
Supplier:  Biorbyt
CAS Number:  
Human recombinant Cystatin C
Catalog # BIRBORB79659-1
Supplier:  Biorbyt
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Protein/peptide type:
    Recombinant
  • Species:
    Human
  • Storage conditions:
    Store at -2 to -8 °C.
  • Protein synonyms:
    AD8
  • Protein/peptide name:
    Cystatin C
  • Purity:
    Greater than 95% pure SDS-PAGE. Protein G Chromatography

Specifications

About this item