Order Entry
Hungary
ContactUsLinkComponent
Human recombinant BMP10
Human recombinant BMP10
Supplier:  Biorbyt
Certificates
Human recombinant BMP10
Supplier:  Biorbyt

Specifications

  • Protein/peptide type:
    Recombinant
  • Species:
    Human
  • Storage conditions:
    The protein is stable at −20 °C. to −70 °C. in a manual defrost freezer.Avoid repeated freeze thaw cycles
  • Protein/peptide name:
    BMP10
  • Purity:
    Greater than 95% by SDS-PAGE gel analysis.
  • Formulation:
    PBS, pH 7.2, sterile filtered
  • Tested applications:
    ELISA , WB , DOT , FuncS

Specifications

Related Information

Frequently Bought Together