Human recombinant IFNA6 (from HEK293 cells)
Supplier: Biorbyt
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage conditions:Store at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
- Protein synonyms:Interferon Alpha6
- UniProtKB:P05013 (Human)
- Protein/peptide name:IFNA6
- Purity:>95% as determined by reducing SDS-PAGE.
- Sequence:SLDCDLPQTHSLGHRRTMMLLAQMRRISLFSCLKDRHDFRFPQEEFDGNQFQKAEAISVLHEVIQ QTFNLFSTKDSSVAWDERLLDKLYTELYQQLNDLEACVMQEVWVGGTPLMNEDSILAVRKYFQRI TLYLTEKKYSPCAWEVVRAEIMRSFSSSRNLQERLRRKEVDHHHHHH
- Formulation:Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, pH 7.4.
- Tested applications:SDS-PAGE
Specifications
Frequently Bought Together