Human recombinant HSPb11 (from E. coli)
: Biorbyt
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:Store at −20 °C, stable for 6 months after receipt. Please minimize freeze-thaw cycles.
- Protein synonyms:Heat Shock Beta11
- UniProtKB:Q9Y547 (Human)
- Protein/peptide name:HSPb11
- Purity:>95% as determined by reducing SDS-PAGE.
- Sequence:MGSSHHHHHHSSGLVPRGSHMRKIDLCLSSEGSEVILATSSDEKHPPENIIDGNPETFWTTTGMF PQEFIICFHKHVRIERLVIQSYFVQTLKIEKSTSKEPVDFEQWIEKDLVHTEGQLQNEEIVAHDG SATYLRFIIVSAFDHFASVHSVSAEGTVVSNLSS
- Formulation:Supplied as a 0.2 um filtered solution of 20mM TrisHCl, 100mM NaCl, 2mM DTT, 10% Glycerol, pH 8.0.
- Tested applications:SDS-PAGE
Frequently Bought Together