Human recombinant MOG (from E. coli)
: Biorbyt
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:Store at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
- Biological activity:Tested for capability to induce EAE in rodents and monkeys
- Protein synonyms:MyelinOligodendrocyte Glycoprotein
- UniProtKB:Q16653 (Human)
- Protein/peptide name:MOG
- Purity:>95% as determined by reducing SDS-PAGE.
- Sequence:MGQFRVIGPRHPIRALVGDEVELPCRISPGKNATGMEVGWYRPPFSRVVHLYRNGKDQDGDQAPE YRGRTELLKDAIGEGKVTLRIRNVRFSDEGGFTCFFRDHSYQEEAAMELKVEDPFYWVSPGHHHH HH
- Formulation:Lyophilized from a 0.2 um filtered solution of 20mM HAc-NaAc, 150mM NaCl, pH 4.5.
- Tested applications:SDS-PAGE , FuncS
Frequently Bought Together