Rendelés felvitel
Hungary
ContactUsLinkComponent
 
Human recombinant LR3IGFI (from E. coli)
 :  Biorbyt
 
Human recombinant LR3IGFI (from E. coli)
 :  Biorbyt

 

  • Konjugáció:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Forrás:
    E. coli
  • Species:
    Human
  • Storage conditions:
    Store at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Biológiai aktivitás:
    ED50 is greater than 200 ng/ml as determined by an IGF binding protein assay.
  • Protein synonyms:
    InsulinLike Growth Factor I
  • UniProtKB:
    P05019 (Human)
  • Protein/peptide name:
    LR3IGFI
  • Tisztaság:
    >95% as determined by reducing SDS-PAGE.
  • Szekvencia:
    GPETLCGAELVDALQFVCGDRGFYFNKPTGYGSSSRRAPQTGIVDECCFRSCDLRRLEMYCAPLK PAKSA
  • Összeállítás:
    Lyophilized from a 0.2 um filtered solution of 300mM NaAc, pH 6.5.
  • Tested applications:
    SDS-PAGE , FuncS

 

Frequently Bought Together