Human recombinant IL8 (from E. coli)
: Biorbyt
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:Store at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
- Biological activity:Recombinant IL8 (379) is fully biologically active when compared to standards.
ED50 is less than 2 ng/ml as determined by its chemotaxis of hCXCR2 transfected mouse BaF/3 cells.
Specific Activity of 5.0 x 105 IU/mg. - Protein synonyms:310C
- UniProtKB:P10145 (Human)
- Protein/peptide name:IL8
- Purity:>95% as determined by reducing SDS-PAGE.
- Sequence:AVLPRSAKELRCQCIKTYSKPFHPKFIKELRVIESGPHCANTEIIVKLSDGRELCLDPKENWVQR VVEKFLKRAENS
- Formulation:Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, pH 7.4.
- Tested applications:SDS-PAGE , FuncS
Frequently Bought Together